Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATTTTCATTCAGTTATGTTTA[A/C]GATGTTTTCAGGCATGCTTGGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609372 MIM: 609373 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM53C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM53C - family with sequence similarity 53 member C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KDM3B - lysine demethylase 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016604.3 | 322 | Intron | NP_057688.2 | |||
XM_005272018.4 | 322 | Intron | XP_005272075.1 | |||
XM_011543488.2 | 322 | Intron | XP_011541790.1 | |||
XM_011543489.2 | 322 | UTR 5 | XP_011541791.1 | |||
XM_017009584.1 | 322 | Intron | XP_016865073.1 |