Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGTGGGGTTAGTCGGGGAGGGG[A/G]TGGACACTGACCTTAAGCCTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612824 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC14L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SEC14L3 - SEC14 like lipid binding 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257378.1 | Intron | NP_001244307.1 | ||||
NM_001257379.1 | Intron | NP_001244308.1 | ||||
NM_001257382.1 | Intron | NP_001244311.1 | ||||
NM_174975.4 | Intron | NP_777635.1 | ||||
XM_011530128.2 | Intron | XP_011528430.1 |