Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606481 MIM: 611651 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5J PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
INPP5J - inositol polyphosphate-5-phosphatase J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002837.2 | 2342 | Intron | NP_001002837.1 | |||
NM_001284285.1 | 2342 | Intron | NP_001271214.1 | |||
NM_001284286.1 | 2342 | Intron | NP_001271215.1 | |||
NM_001284287.1 | 2342 | Intron | NP_001271216.1 | |||
NM_001284288.1 | 2342 | Intron | NP_001271217.1 | |||
NM_001284289.1 | 2342 | Intron | NP_001271218.1 | |||
XM_011530143.1 | 2342 | Intron | XP_011528445.1 | |||
XM_017028771.1 | 2342 | Intron | XP_016884260.1 | |||
XM_017028772.1 | 2342 | Intron | XP_016884261.1 | |||
XM_017028773.1 | 2342 | Intron | XP_016884262.1 |
PLA2G3 - phospholipase A2 group III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015715.4 | 2342 | UTR 3 | NP_056530.2 | |||
XM_011530204.1 | 2342 | UTR 3 | XP_011528506.1 | |||
XM_011530205.1 | 2342 | UTR 3 | XP_011528507.1 |
Set Membership: |
HapMap Validated |