Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147811 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1R2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL1R2 - interleukin 1 receptor type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261419.1 | 270 | Intron | NP_001248348.1 | |||
NM_004633.3 | 270 | Intron | NP_004624.1 | |||
XM_006712734.3 | 270 | Intron | XP_006712797.1 | |||
XM_006712736.3 | 270 | Intron | XP_006712799.1 | |||
XM_011511801.2 | 270 | Intron | XP_011510103.1 | |||
XM_011511803.2 | 270 | UTR 5 | XP_011510105.1 | |||
XM_011511804.2 | 270 | Intron | XP_011510106.1 | |||
XM_011511805.2 | 270 | Intron | XP_011510107.1 | |||
XM_011511807.1 | 270 | Intron | XP_011510109.1 | |||
XM_011511808.2 | 270 | Intron | XP_011510110.1 | |||
XM_017004889.1 | 270 | Intron | XP_016860378.1 |
Set Membership: |
HapMap |