Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGTAATAGCCAACTTGGTTGCA[A/G]CATTTAAGGTCATGCCTGTCTAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608444 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT2E PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KMT2E - lysine methyltransferase 2E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018682.3 | Intron | NP_061152.3 | ||||
NM_182931.2 | Intron | NP_891847.1 | ||||
XM_005250493.1 | Intron | XP_005250550.1 | ||||
XM_011516400.1 | Intron | XP_011514702.1 | ||||
XM_017012435.1 | Intron | XP_016867924.1 |
KMT2E-AS1 - KMT2E antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |