Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605678 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLXIPL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MLXIPL - MLX interacting protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032951.2 | 2859 | UTR 3 | NP_116569.1 | |||
NM_032952.2 | 2859 | UTR 3 | NP_116570.1 | |||
NM_032953.2 | 2859 | UTR 3 | NP_116571.1 | |||
NM_032954.2 | 2859 | UTR 3 | NP_116572.1 | |||
XM_011516277.1 | 2859 | UTR 3 | XP_011514579.1 | |||
XM_011516278.1 | 2859 | UTR 3 | XP_011514580.1 | |||
XM_011516279.1 | 2859 | UTR 3 | XP_011514581.1 | |||
XM_011516281.2 | 2859 | UTR 3 | XP_011514583.1 | |||
XM_017012263.1 | 2859 | UTR 3 | XP_016867752.1 |
Set Membership: |
HapMap |