Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATTCTCCTCTGCTGGACAAAGA[A/G]TGACTGAGTTGGAACCTCAGAGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102681 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADD2 - adducin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185054.1 | 3505 | UTR 3 | NP_001171983.1 | |||
NM_001185055.1 | 3505 | Intron | NP_001171984.1 | |||
NM_001617.3 | 3505 | UTR 3 | NP_001608.1 | |||
NM_017482.3 | 3505 | Intron | NP_059516.2 | |||
NM_017488.3 | 3505 | UTR 3 | NP_059522.1 | |||
XM_011532502.2 | 3505 | UTR 3 | XP_011530804.1 |
Set Membership: |
HapMap |