Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAAACAGACAGGAAACACACCAT[A/G]CATATTCAGAGGGAAAGTCAAATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609907 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEMA3D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEMA3D - semaphorin 3D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152754.2 | 4398 | UTR 3 | NP_689967.2 | |||
XM_011515960.1 | 4398 | UTR 3 | XP_011514262.1 | |||
XM_011515961.2 | 4398 | UTR 3 | XP_011514263.1 | |||
XM_017011873.1 | 4398 | UTR 3 | XP_016867362.1 |
Set Membership: |
HapMap |