Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAGACCAGGAAAGCCAGGCCCG[C/T]GGGGAGGGGTAAGGAGAGAGGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608171 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA2D4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CACNA2D4 - calcium voltage-gated channel auxiliary subunit alpha2delta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172364.4 | Intron | NP_758952.4 | ||||
XM_011521041.2 | Intron | XP_011519343.1 |
LRTM2 - leucine rich repeats and transmembrane domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039029.2 | Intron | NP_001034118.1 | ||||
NM_001163925.1 | Intron | NP_001157397.1 | ||||
NM_001163926.1 | Intron | NP_001157398.1 | ||||
XM_011521015.1 | Intron | XP_011519317.1 | ||||
XM_017019848.1 | Intron | XP_016875337.1 |
Set Membership: |
HapMap |