Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAGGACAAGATGGGAAAACTGC[A/T]TTCTAATTCACCTAGGGCCCAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607946 MIM: 608171 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADIPOR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADIPOR2 - adiponectin receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CACNA2D4 - calcium voltage-gated channel auxiliary subunit alpha2delta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172364.4 | Intron | NP_758952.4 | ||||
XM_011521041.2 | Intron | XP_011519343.1 |
Set Membership: |
HapMap Validated |