Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATATGTTAAATGCAAAACAGAA[C/T]TCTTGCTAGCTAAACACAGTAGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615004 MIM: 605224 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRIT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LRIT3 - leucine rich repeat, Ig-like and transmembrane domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198506.4 | Intron | NP_940908.3 | ||||
XM_017008167.1 | Intron | XP_016863656.1 | ||||
XM_017008168.1 | Intron | XP_016863657.1 |
RRH - retinal pigment epithelium-derived rhodopsin homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |