Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614178 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRACR2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRACR2A - calcium release activated channel regulator 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144958.1 | Intron | NP_001138430.1 | ||||
NM_032680.3 | Intron | NP_116069.1 | ||||
XM_006719021.3 | Intron | XP_006719084.1 | ||||
XM_011521034.2 | Intron | XP_011519336.1 | ||||
XM_011521036.2 | Intron | XP_011519338.1 | ||||
XM_011521037.2 | Intron | XP_011519339.1 | ||||
XM_011521038.2 | Intron | XP_011519340.1 |