Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCCCAGCCCAACTCCCATTGTC[A/G]TAAAAAACATTGACCCTGATTAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604689 MIM: 603834 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKAP3 - A-kinase anchoring protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFA9 - NADH:ubiquinone oxidoreductase subunit A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005002.4 | Intron | NP_004993.1 |
Set Membership: |
HapMap |