Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCACAGGCGGGGCTGGGTGGGGC[A/G]TGGGCTGGCAAGGAAGTGAACCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601719 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBX4 - T-box 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321120.1 | Intron | NP_001308049.1 | ||||
NM_018488.3 | Intron | NP_060958.2 | ||||
XM_011525490.2 | Intron | XP_011523792.1 | ||||
XM_011525491.2 | Intron | XP_011523793.1 | ||||
XM_011525495.2 | Intron | XP_011523797.1 |