Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAAGAATATTTCATAGGCTGTAC[C/T]ATGTACTTTGCATCATATCAGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605947 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIGL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PIGL - phosphatidylinositol glycan anchor biosynthesis class L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004278.3 | Intron | NP_004269.1 | ||||
XM_011524080.2 | Intron | XP_011522382.1 | ||||
XM_017025349.1 | Intron | XP_016880838.1 | ||||
XM_017025350.1 | Intron | XP_016880839.1 | ||||
XM_017025351.1 | Intron | XP_016880840.1 | ||||
XM_017025352.1 | Intron | XP_016880841.1 | ||||
XM_017025353.1 | Intron | XP_016880842.1 | ||||
XM_017025354.1 | Intron | XP_016880843.1 | ||||
XM_017025355.1 | Intron | XP_016880844.1 | ||||
XM_017025356.1 | Intron | XP_016880845.1 |
Set Membership: |
HapMap Validated |