Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAATATTCATCAACATCATCAC[C/T]ATCATCAGCTGTGGGCTGAGGTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609603 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYGN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRYGN - crystallin gamma N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308292.1 | 1096 | UTR 3 | NP_001295221.1 | |||
NM_144727.2 | 1096 | UTR 3 | NP_653328.1 | |||
XM_005249952.3 | 1096 | Intron | XP_005250009.1 | |||
XM_011515836.1 | 1096 | Intron | XP_011514138.1 | |||
XM_011515837.2 | 1096 | Intron | XP_011514139.1 | |||
XM_017011773.1 | 1096 | Intron | XP_016867262.1 |
MIR3907 - microRNA 3907 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |