Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604086 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF155 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF155 - zinc finger protein 155 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF221 - zinc finger protein 221 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297588.1 | Intron | NP_001284517.1 | ||||
NM_001297589.1 | Intron | NP_001284518.1 | ||||
NM_013359.2 | Intron | NP_037491.2 | ||||
XM_017027232.1 | Intron | XP_016882721.1 | ||||
XM_017027233.1 | Intron | XP_016882722.1 |
Set Membership: |
HapMap |