Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176636 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRIM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PRIM2 - primase (DNA) subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000947.4 | Intron | NP_000938.2 | ||||
NM_001282487.1 | Intron | NP_001269416.1 | ||||
NM_001282488.1 | Intron | NP_001269417.1 | ||||
XM_011514731.2 | Intron | XP_011513033.1 | ||||
XM_017011004.1 | Intron | XP_016866493.1 | ||||
XM_017011005.1 | Intron | XP_016866494.1 | ||||
XM_017011006.1 | Intron | XP_016866495.1 | ||||
XM_017011007.1 | Intron | XP_016866496.1 |
Set Membership: |
HapMap Validated |