Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606580 MIM: 601703 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OPA3 - optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017989.2 | 2189 | Intron | NP_001017989.2 | |||
NM_025136.3 | 2189 | Intron | NP_079412.1 | |||
XM_006723403.3 | 2189 | Intron | XP_006723466.1 |
VASP - vasodilator-stimulated phosphoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003370.3 | 2189 | UTR 3 | NP_003361.1 | |||
XM_005259199.1 | 2189 | UTR 3 | XP_005259256.1 | |||
XM_005259200.1 | 2189 | UTR 3 | XP_005259257.1 | |||
XM_017027200.1 | 2189 | UTR 3 | XP_016882689.1 |
Set Membership: |
HapMap |