Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCCCATCCCCATCATAATGGAT[G/T]TCTAACTTCAAGGAGCCAGAGAACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPINK14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SPINK14 - serine peptidase inhibitor, Kazal type 14 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001325.2 | Intron | NP_001001325.1 | ||||
XM_017009466.1 | Intron | XP_016864955.1 | ||||
XM_017009467.1 | Intron | XP_016864956.1 | ||||
XM_017009468.1 | Intron | XP_016864957.1 | ||||
XM_017009469.1 | Intron | XP_016864958.1 |
Set Membership: |
HapMap |