Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTGCAACCTAATAAGGTTCCAT[C/T]CATCCAGTGACTCGTTCCTCATATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF582-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF582-AS1 - ZNF582 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF583 - zinc finger protein 583 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159860.1 | Intron | NP_001153332.1 | ||||
NM_001159861.1 | Intron | NP_001153333.1 | ||||
NM_152478.2 | Intron | NP_689691.2 | ||||
XM_005258558.1 | Intron | XP_005258615.1 | ||||
XM_011526517.2 | Intron | XP_011524819.1 | ||||
XM_017026346.1 | Intron | XP_016881835.1 | ||||
XM_017026347.1 | Intron | XP_016881836.1 | ||||
XM_017026348.1 | Intron | XP_016881837.1 | ||||
XM_017026349.1 | Intron | XP_016881838.1 | ||||
XM_017026350.1 | Intron | XP_016881839.1 | ||||
XM_017026351.1 | Intron | XP_016881840.1 | ||||
XM_017026352.1 | Intron | XP_016881841.1 | ||||
XM_017026353.1 | Intron | XP_016881842.1 |