Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAATTTTAAGGTAGTTTGTTGTA[A/T]GCTGGAAGCAATTCAATTGTTTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114204 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA2D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CACNA2D1 - calcium voltage-gated channel auxiliary subunit alpha2delta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000722.3 | 6026 | UTR 3 | NP_000713.2 | |||
NM_001302890.1 | 6026 | Intron | NP_001289819.1 | |||
XM_005250570.2 | 6026 | UTR 3 | XP_005250627.1 | |||
XM_005250572.2 | 6026 | UTR 3 | XP_005250629.1 | |||
XM_005250573.2 | 6026 | UTR 3 | XP_005250630.1 | |||
XM_005250574.2 | 6026 | UTR 3 | XP_005250631.1 | |||
XM_006716118.2 | 6026 | UTR 3 | XP_006716181.1 | |||
XM_006716119.3 | 6026 | UTR 3 | XP_006716182.1 | |||
XM_006716120.3 | 6026 | UTR 3 | XP_006716183.1 | |||
XM_006716121.3 | 6026 | UTR 3 | XP_006716184.1 | |||
XM_011516570.2 | 6026 | Intron | XP_011514872.1 | |||
XM_011516571.2 | 6026 | UTR 3 | XP_011514873.1 | |||
XM_011516572.2 | 6026 | UTR 3 | XP_011514874.1 | |||
XM_017012588.1 | 6026 | UTR 3 | XP_016868077.1 |
Set Membership: |
HapMap Validated |