Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604511 MIM: 614906 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CBX1 - chromobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX11 - sorting nexin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013323.2 | Intron | NP_037455.2 | ||||
NM_152244.1 | Intron | NP_689450.1 | ||||
XM_005257260.3 | Intron | XP_005257317.1 | ||||
XM_005257261.3 | Intron | XP_005257318.1 | ||||
XM_005257262.3 | Intron | XP_005257319.1 | ||||
XM_011524697.2 | Intron | XP_011522999.1 |