Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATTATAGCTATGTCTGAGAAA[A/T]GCTATCTCCCATATAAGGCTTAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615078 MIM: 600537 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLT1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GOLT1B - golgi transport 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016072.4 | Intron | NP_057156.1 | ||||
XM_017019384.1 | Intron | XP_016874873.1 |
RECQL - RecQ like helicase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |