Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185490 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC149 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC149 - coiled-coil domain containing 149 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005248210.3 | 3399 | UTR 3 | XP_005248267.1 | |||
XM_011513906.2 | 3399 | UTR 3 | XP_011512208.1 | |||
XM_011513907.2 | 3399 | UTR 3 | XP_011512209.1 | |||
XM_011513908.1 | 3399 | UTR 3 | XP_011512210.1 | |||
XM_017008827.1 | 3399 | UTR 3 | XP_016864316.1 | |||
XM_017008828.1 | 3399 | UTR 3 | XP_016864317.1 |
SOD3 - superoxide dismutase 3, extracellular | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |