Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGTGTTGTAGAAACTTGCAAAAA[C/T]TTGTCAGCAATGGCACTATTTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601762 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CASP10 - caspase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206524.1 | Intron | NP_001193453.1 | ||||
NM_001206542.1 | Intron | NP_001193471.1 | ||||
NM_001230.4 | Intron | NP_001221.2 | ||||
NM_001306083.1 | Intron | NP_001293012.1 | ||||
NM_032974.4 | Intron | NP_116756.2 | ||||
NM_032976.3 | Intron | NP_116758.1 | ||||
NM_032977.3 | Intron | NP_116759.2 | ||||
XM_005246907.2 | Intron | XP_005246964.1 | ||||
XM_006712796.3 | Intron | XP_006712859.1 | ||||
XM_011511990.2 | Intron | XP_011510292.1 |
Set Membership: |
HapMap Validated |