Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 152310 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFPI PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TFPI - tissue factor pathway inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005246818.1 | Intron | XP_005246875.1 | ||||
XM_005246819.1 | Intron | XP_005246876.1 | ||||
XM_006712720.3 | Intron | XP_006712783.1 | ||||
XM_011511707.2 | Intron | XP_011510009.1 | ||||
XM_011511708.1 | Intron | XP_011510010.1 | ||||
XM_011511709.2 | Intron | XP_011510011.1 |
Set Membership: |
HapMap |