Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTACACCCTTGTTGGCCTGTTGCT[A/G]ACATCATTCAGTTTTCCTCACTGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300681 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DOCK11 - dedicator of cytokinesis 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144658.3 | Intron | NP_653259.3 | ||||
XM_005262368.3 | Intron | XP_005262425.1 | ||||
XM_005262369.4 | Intron | XP_005262426.1 | ||||
XM_011531276.2 | Intron | XP_011529578.1 | ||||
XM_011531278.2 | Intron | XP_011529580.1 |
Set Membership: |
HapMap |