Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602449 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKAP1 - A-kinase anchoring protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242902.1 | Intron | NP_001229831.1 | ||||
NM_001242903.1 | Intron | NP_001229832.1 | ||||
NM_003488.3 | Intron | NP_003479.1 | ||||
XM_005257707.2 | Intron | XP_005257764.1 | ||||
XM_005257709.2 | Intron | XP_005257766.1 | ||||
XM_017025189.1 | Intron | XP_016880678.1 | ||||
XM_017025190.1 | Intron | XP_016880679.1 | ||||
XM_017025191.1 | Intron | XP_016880680.1 |
Set Membership: |
HapMap |