Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194545 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF71 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF71 - zinc finger protein 71 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021216.4 | 1797 | Intron | NP_067039.1 | |||
XM_011527193.2 | 1797 | Intron | XP_011525495.1 | |||
XM_017027078.1 | 1797 | Intron | XP_016882567.1 | |||
XM_017027079.1 | 1797 | UTR 5 | XP_016882568.1 | |||
XM_017027080.1 | 1797 | Intron | XP_016882569.1 |
Set Membership: |
HapMap |