Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCATTTCTCCATGGATGTTTAG[C/T]TCTTTCTTTTTAGTGGAAAATGGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616147 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDK15 - cyclin dependent kinase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261435.1 | Intron | NP_001248364.1 | ||||
NM_001261436.1 | Intron | NP_001248365.1 | ||||
NM_139158.2 | Intron | NP_631897.1 | ||||
XM_005246781.3 | Intron | XP_005246838.1 | ||||
XM_005246782.4 | Intron | XP_005246839.1 | ||||
XM_011511650.2 | Intron | XP_011509952.1 | ||||
XM_011511652.1 | Intron | XP_011509954.1 | ||||
XM_011511654.2 | Intron | XP_011509956.1 | ||||
XM_011511655.2 | Intron | XP_011509957.1 | ||||
XM_011511656.2 | Intron | XP_011509958.1 | ||||
XM_011511657.2 | Intron | XP_011509959.1 |
Set Membership: |
HapMap |