Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608111 MIM: 602169 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FANCL - Fanconi anemia complementation group L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114636.1 | 1447 | Intron | NP_001108108.1 | |||
NM_018062.3 | 1447 | Intron | NP_060532.2 | |||
XM_005264395.3 | 1447 | UTR 3 | XP_005264452.1 | |||
XM_005264397.4 | 1447 | Intron | XP_005264454.1 | |||
XM_011532939.2 | 1447 | UTR 3 | XP_011531241.1 | |||
XM_011532940.2 | 1447 | UTR 3 | XP_011531242.1 | |||
XM_011532941.1 | 1447 | Intron | XP_011531243.1 | |||
XM_011532942.2 | 1447 | UTR 3 | XP_011531244.1 | |||
XM_011532944.2 | 1447 | UTR 3 | XP_011531246.1 | |||
XM_011532945.2 | 1447 | UTR 3 | XP_011531247.1 | |||
XM_017004414.1 | 1447 | Intron | XP_016859903.1 | |||
XM_017004415.1 | 1447 | Intron | XP_016859904.1 | |||
XM_017004416.1 | 1447 | UTR 3 | XP_016859905.1 |
Set Membership: |
HapMap |