Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST6GAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST6GAL1 - ST6 beta-galactoside alpha-2,6-sialyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003032.2 | Intron | NP_003023.1 | ||||
NM_173216.2 | Intron | NP_775323.1 | ||||
NM_173217.2 | Intron | NP_775324.1 | ||||
XM_005247719.1 | Intron | XP_005247776.1 | ||||
XM_005247720.1 | Intron | XP_005247777.1 | ||||
XM_006713734.1 | Intron | XP_006713797.1 | ||||
XM_011513085.1 | Intron | XP_011511387.1 | ||||
XM_011513086.1 | Intron | XP_011511388.1 | ||||
XM_017007065.1 | Intron | XP_016862554.1 | ||||
XM_017007066.1 | Intron | XP_016862555.1 | ||||
XM_017007067.1 | Intron | XP_016862556.1 | ||||
XM_017007068.1 | Intron | XP_016862557.1 |