Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613894 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPY19L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DPY19L3 - dpy-19 like 3 (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172774.1 | Intron | NP_001166245.1 | ||||
NM_207325.2 | Intron | NP_997208.2 | ||||
XM_006723045.1 | Intron | XP_006723108.1 | ||||
XM_011526526.2 | Intron | XP_011524828.1 | ||||
XM_017026366.1 | Intron | XP_016881855.1 | ||||
XM_017026367.1 | Intron | XP_016881856.1 |
LOC400684 - uncharacterized LOC400684 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |