Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAAGCATGTTAGCCAAGGGAAG[G/T]TAGGGCCACAACTGTCCAGAGCCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604768 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF254 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF254 - zinc finger protein 254 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278661.1 | Intron | NP_001265590.1 | ||||
NM_001278662.1 | Intron | NP_001265591.1 | ||||
NM_001278663.1 | Intron | NP_001265592.1 | ||||
NM_001278664.1 | Intron | NP_001265593.1 | ||||
NM_001278665.1 | Intron | NP_001265594.1 | ||||
NM_001278677.1 | Intron | NP_001265606.1 | ||||
NM_001278678.1 | Intron | NP_001265607.1 | ||||
NM_203282.3 | Intron | NP_975011.3 | ||||
XM_011528443.1 | Intron | XP_011526745.1 | ||||
XM_011528444.1 | Intron | XP_011526746.1 | ||||
XM_011528448.2 | Intron | XP_011526750.1 | ||||
XM_017027514.1 | Intron | XP_016883003.1 | ||||
XM_017027515.1 | Intron | XP_016883004.1 | ||||
XM_017027516.1 | Intron | XP_016883005.1 | ||||
XM_017027517.1 | Intron | XP_016883006.1 | ||||
XM_017027518.1 | Intron | XP_016883007.1 | ||||
XM_017027519.1 | Intron | XP_016883008.1 |
Set Membership: |
HapMap |