Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 141180 MIM: 171891 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCKAP1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NCKAP1L - NCK associated protein 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE1B - phosphodiesterase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000924.3 | Intron | NP_000915.1 | ||||
NM_001165975.2 | Intron | NP_001159447.1 | ||||
NM_001288768.1 | Intron | NP_001275697.1 | ||||
NM_001288769.1 | Intron | NP_001275698.1 | ||||
NM_001315534.1 | Intron | NP_001302463.1 | ||||
NM_001315535.1 | Intron | NP_001302464.1 | ||||
XM_011538456.1 | Intron | XP_011536758.1 | ||||
XM_017019432.1 | Intron | XP_016874921.1 | ||||
XM_017019433.1 | Intron | XP_016874922.1 |
Set Membership: |
HapMap Validated |