Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607218 MIM: 610032 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IRF5 - interferon regulatory factor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNPO3 - transportin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191028.2 | 3921 | UTR 3 | NP_001177957.2 | |||
NM_012470.3 | 3921 | UTR 3 | NP_036602.1 | |||
XM_011515989.2 | 3921 | Intron | XP_011514291.1 |
Set Membership: |
HapMap Validated |