Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614539 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HELB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HELB - DNA helicase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033647.4 | 86 | Silent Mutation | CGC,CGT | R,R 9 | NP_387467.2 | |
XM_005269234.2 | 86 | Silent Mutation | CGC,CGT | R,R 9 | XP_005269291.1 | |
XM_017020230.1 | 86 | Silent Mutation | CGC,CGT | R,R 9 | XP_016875719.1 |
Set Membership: |
HapMap Validated |