Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAAGACCCAATCTGGATCAGCC[C/T]CTTTCCACATCTGCATGCAGTTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608481 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC26A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC26A9 - solute carrier family 26 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052934.3 | 3475 | UTR 3 | NP_443166.1 | |||
NM_134325.2 | 3475 | UTR 3 | NP_599152.2 | |||
XM_011509121.2 | 3475 | UTR 3 | XP_011507423.1 | |||
XM_011509122.2 | 3475 | UTR 3 | XP_011507424.1 | |||
XM_011509124.2 | 3475 | Intron | XP_011507426.1 |