Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAATTTGGCACTTCGGGTGCTG[G/T]GCACGGTGAATCCCAGATAACATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608481 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC26A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC26A9 - solute carrier family 26 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052934.3 | 3997 | UTR 3 | NP_443166.1 | |||
NM_134325.2 | 3997 | UTR 3 | NP_599152.2 | |||
XM_011509121.2 | 3997 | UTR 3 | XP_011507423.1 | |||
XM_011509122.2 | 3997 | UTR 3 | XP_011507424.1 | |||
XM_011509124.2 | 3997 | Intron | XP_011507426.1 |
Set Membership: |
HapMap |