Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615569 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFXN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SFXN1 - sideroflexin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322977.1 | Intron | NP_001309906.1 | ||||
NM_001322978.1 | Intron | NP_001309907.1 | ||||
NM_001322980.1 | Intron | NP_001309909.1 | ||||
NM_001322981.1 | Intron | NP_001309910.1 | ||||
NM_001322982.1 | Intron | NP_001309911.1 | ||||
NM_001322983.1 | Intron | NP_001309912.1 | ||||
NM_022754.6 | Intron | NP_073591.2 |
Set Membership: |
HapMap |