Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACTGGAACTGGGCATGAATATA[A/C]TTCTAAAATGAGATTCCTGAAGTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614161 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRDM5 - PR domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300823.1 | 3515 | UTR 3 | NP_001287752.1 | |||
NM_001300824.1 | 3515 | UTR 3 | NP_001287753.1 | |||
NM_018699.3 | 3515 | UTR 3 | NP_061169.2 | |||
XM_005262708.3 | 3515 | Intron | XP_005262765.1 | |||
XM_011531562.2 | 3515 | UTR 3 | XP_011529864.1 | |||
XM_011531563.2 | 3515 | UTR 3 | XP_011529865.1 | |||
XM_011531564.2 | 3515 | UTR 3 | XP_011529866.1 | |||
XM_011531565.2 | 3515 | Intron | XP_011529867.1 | |||
XM_011531566.2 | 3515 | Intron | XP_011529868.1 | |||
XM_011531567.2 | 3515 | Intron | XP_011529869.1 | |||
XM_011531568.2 | 3515 | Intron | XP_011529870.1 | |||
XM_011531569.2 | 3515 | Intron | XP_011529871.1 | |||
XM_011531570.2 | 3515 | UTR 3 | XP_011529872.1 | |||
XM_017007668.1 | 3515 | UTR 3 | XP_016863157.1 | |||
XM_017007669.1 | 3515 | Intron | XP_016863158.1 | |||
XM_017007670.1 | 3515 | Intron | XP_016863159.1 | |||
XM_017007671.1 | 3515 | UTR 3 | XP_016863160.1 |
Set Membership: |
HapMap |