Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTAGTCTCCTACGCTTCTTTTC[C/T]AGTACATGAAACATTACCATATGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602325 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4G2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EIF4G2 - eukaryotic translation initiation factor 4 gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042559.2 | Intron | NP_001036024.3 | ||||
NM_001172705.1 | Intron | NP_001166176.1 | ||||
NM_001418.3 | Intron | NP_001409.3 |
LOC101928053 - uncharacterized LOC101928053 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD97 - small nucleolar RNA, C/D box 97 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |