Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATCTCTGCTTCTATCAAGGGAAG[C/G]CCATTATGTTCTATGCACATATATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607296 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1G1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP6V1G1 - ATPase H+ transporting V1 subunit G1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM268 - transmembrane protein 268 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153045.3 | Intron | NP_694590.2 | ||||
XM_006716997.1 | Intron | XP_006717060.1 | ||||
XM_011518350.1 | Intron | XP_011516652.1 | ||||
XM_011518351.2 | Intron | XP_011516653.1 | ||||
XM_011518352.1 | Intron | XP_011516654.1 | ||||
XM_011518353.1 | Intron | XP_011516655.1 | ||||
XM_011518354.1 | Intron | XP_011516656.1 | ||||
XM_011518355.1 | Intron | XP_011516657.1 | ||||
XM_017014429.1 | Intron | XP_016869918.1 | ||||
XM_017014430.1 | Intron | XP_016869919.1 |
Set Membership: |
HapMap |