Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616518 MIM: 611023 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC38A6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC38A6 - solute carrier family 38 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172702.1 | Intron | NP_001166173.1 | ||||
NM_153811.2 | Intron | NP_722518.2 | ||||
XM_006720049.3 | Intron | XP_006720112.1 | ||||
XM_006720050.1 | Intron | XP_006720113.1 | ||||
XM_011536469.2 | Intron | XP_011534771.1 | ||||
XM_017021020.1 | Intron | XP_016876509.1 | ||||
XM_017021021.1 | Intron | XP_016876510.1 | ||||
XM_017021022.1 | Intron | XP_016876511.1 | ||||
XM_017021023.1 | Intron | XP_016876512.1 | ||||
XM_017021024.1 | Intron | XP_016876513.1 | ||||
XM_017021025.1 | Intron | XP_016876514.1 | ||||
XM_017021026.1 | Intron | XP_016876515.1 | ||||
XM_017021027.1 | Intron | XP_016876516.1 | ||||
XM_017021028.1 | Intron | XP_016876517.1 |
TRMT5 - tRNA methyltransferase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |