Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607697 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SBF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SBF2 - SET binding factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030962.3 | 7227 | UTR 3 | NP_112224.1 | |||
XM_005253154.4 | 7227 | UTR 3 | XP_005253211.1 | |||
XM_005253155.4 | 7227 | UTR 3 | XP_005253212.1 | |||
XM_011520394.2 | 7227 | UTR 3 | XP_011518696.1 | |||
XM_011520395.2 | 7227 | Intron | XP_011518697.1 | |||
XM_011520396.2 | 7227 | Intron | XP_011518698.1 | |||
XM_017018372.1 | 7227 | UTR 3 | XP_016873861.1 | |||
XM_017018373.1 | 7227 | UTR 3 | XP_016873862.1 | |||
XM_017018374.1 | 7227 | UTR 3 | XP_016873863.1 | |||
XM_017018375.1 | 7227 | UTR 3 | XP_016873864.1 | |||
XM_017018376.1 | 7227 | Intron | XP_016873865.1 | |||
XM_017018377.1 | 7227 | Intron | XP_016873866.1 |
SBF2-AS1 - SBF2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |