Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613131 MIM: 613132 | ||||||||||||||||||||
Literature Links: |
CDC20B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC20B - cell division cycle 20B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145734.2 | Intron | NP_001139206.2 | ||||
NM_001170402.1 | Intron | NP_001163873.1 | ||||
NM_152623.2 | Intron | NP_689836.2 | ||||
XM_011543218.2 | Intron | XP_011541520.1 |
GPX8 - glutathione peroxidase 8 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR449A - microRNA 449a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR449B - microRNA 449b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR449C - microRNA 449c | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |