Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGACTGAGCACTCTGAGGAGGA[C/G]GCTGACTCAGGCTGACCCTTCCCGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615333 | |||||||||||||||||||||||
Literature Links: |
B3GNT5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
B3GNT5 - UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032047.4 | 562 | Intron | NP_114436.1 | |||
XM_005247823.4 | 562 | Intron | XP_005247880.1 | |||
XM_005247824.3 | 562 | Intron | XP_005247881.1 | |||
XM_005247825.3 | 562 | Intron | XP_005247882.1 | |||
XM_011513225.2 | 562 | Intron | XP_011511527.1 | |||
XM_011513226.2 | 562 | Intron | XP_011511528.1 | |||
XM_011513227.2 | 562 | Intron | XP_011511529.1 | |||
XM_011513228.2 | 562 | Intron | XP_011511530.1 | |||
XM_011513229.2 | 562 | Intron | XP_011511531.1 | |||
XM_017007309.1 | 562 | UTR 5 | XP_016862798.1 |
MCF2L2 - MCF.2 cell line derived transforming sequence-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015078.3 | 562 | Intron | NP_055893.3 | |||
XM_011512585.2 | 562 | Intron | XP_011510887.1 | |||
XM_011512586.2 | 562 | Intron | XP_011510888.1 | |||
XM_017005943.1 | 562 | Intron | XP_016861432.1 | |||
XM_017005944.1 | 562 | Intron | XP_016861433.1 | |||
XM_017005945.1 | 562 | Intron | XP_016861434.1 | |||
XM_017005946.1 | 562 | Intron | XP_016861435.1 |