Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615333 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GNT5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
B3GNT5 - UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032047.4 | 172 | Intron | NP_114436.1 | |||
XM_005247823.4 | 172 | Intron | XP_005247880.1 | |||
XM_005247824.3 | 172 | Intron | XP_005247881.1 | |||
XM_005247825.3 | 172 | Intron | XP_005247882.1 | |||
XM_011513225.2 | 172 | Intron | XP_011511527.1 | |||
XM_011513226.2 | 172 | Intron | XP_011511528.1 | |||
XM_011513227.2 | 172 | Intron | XP_011511529.1 | |||
XM_011513228.2 | 172 | Intron | XP_011511530.1 | |||
XM_011513229.2 | 172 | Intron | XP_011511531.1 | |||
XM_017007309.1 | 172 | UTR 5 | XP_016862798.1 |
MCF2L2 - MCF.2 cell line derived transforming sequence-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015078.3 | 172 | Intron | NP_055893.3 | |||
XM_011512585.2 | 172 | Intron | XP_011510887.1 | |||
XM_011512586.2 | 172 | Intron | XP_011510888.1 | |||
XM_017005943.1 | 172 | Intron | XP_016861432.1 | |||
XM_017005944.1 | 172 | Intron | XP_016861433.1 | |||
XM_017005945.1 | 172 | Intron | XP_016861434.1 | |||
XM_017005946.1 | 172 | Intron | XP_016861435.1 |
Set Membership: |
HapMap |