Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTAGGGCCCCTCTGGCCTGGGC[A/G]GTTCCAGGCTCCTTCTCTGCAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF879 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF879 - zinc finger protein 879 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136116.1 | 802 | Intron | NP_001129588.1 | |||
XM_005265908.3 | 802 | UTR 5 | XP_005265965.1 | |||
XM_011534550.2 | 802 | Intron | XP_011532852.1 | |||
XM_011534551.2 | 802 | Intron | XP_011532853.1 |